N-Methyl-4-boronobenzolsulfonamid pinacol ester

Lieferant: COMBI-BLOCKS

Synonyme: N-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide

PN-2610-1G PN-2610-250M
COBBPN-2610-1GEA 298 EUR
COBBPN-2610-1G COBBPN-2610-250M
N-Methyl-4-boronobenzolsulfonamid pinacol ester
N-Methyl-4-boronobenzolsulfonamid pinacol ester
Formel: C₁₃H₂₀BNO₄S
Molecular Weight: 297,1800 g/mol
MDL: MFCD06657895
CAS-Nummer: 1073353-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR