1-Boc-4-piperidinessigsäure

Lieferant: COMBI-BLOCKS

Synonyme: [1-(tert-Butoxycarbonyl)piperidin-4-yl]acetic acid, 2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)acetic acid, 1-(tert-Butoxycarbonyl)-4-piperidylacetic acid

HC-6319-25G HC-6319-5G HC-6319-1G
COBBHC-6319-25GEA 169 EUR
COBBHC-6319-25G COBBHC-6319-5G COBBHC-6319-1G
1-Boc-4-piperidinessigsäure
1-Boc-4-piperidinessigsäure
Formel: C₁₂H₂₁NO₄
Molecular Weight: 243,3000 g/mol
MDL: MFCD00800239
CAS-Nummer: 157688-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR