1-Boc-4-piperidinessigsäure

Lieferant: FLUOROCHEM

Synonyme: [1-(tert-Butoxycarbonyl)piperidin-4-yl]acetic acid, 2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)acetic acid, 1-(tert-Butoxycarbonyl)-4-piperidylacetic acid

011563-1G 011563-5G
FLUO011563-1GEA 31.1 EUR
FLUO011563-1G FLUO011563-5G
1-Boc-4-piperidinessigsäure
1-Boc-4-piperidinessigsäure
Formel: C₁₂H₂₁NO₄
Molecular Weight: 243.3000 g/mol
MDL: MFCD00800239
CAS-Nummer: 157688-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR