(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: (3S)-1,2,3,4-Tetrahydro-3-isoquinolinecarbocylic acid, (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

L14422.03 L14422.06 L14422.14
L14422.03EA 25.7 EUR
L14422.03 L14422.06 L14422.14
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure ≥97%
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure

Purity: 97% CAS No.: 74163-81-8 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Melting Point: 300° dec. Beilstein Reference: 4842199

Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,20 g/mol
Schmelzpunkt: >300 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00144533
CAS-Nummer: 74163-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR