(±)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure

Lieferant: Thermo Scientific

Synonyme: 1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, H-DL-Tic-OH

SB01676.1 SB01676.10
MAYBSB01676.1EA 0 EUR
MAYBSB01676.1 MAYBSB01676.10
(±)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
(±)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177.1998 g/mol
MDL: MFCD00191496
CAS-Nummer: 67123-97-1
EINECS: 266-580-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR