2-Brom-5-(4-morpholino)pyridin
Lieferant: COMBI-BLOCKS
Synonyme:
4-(5-Bromopyridin-2-yl)morpholine, 4-(5-Bromo-2-pyridyl)morpholine, 4-(5-Bromo-pyridin-2-yl)-morpholine
PY-7768-25G
PY-7768-5G
PY-7768-1G
COBBPY-7768-25GEA
404
EUR
COBBPY-7768-25G
COBBPY-7768-5G
COBBPY-7768-1G
2-Brom-5-(4-morpholino)pyridin
2-Brom-5-(4-morpholino)pyridin
Formel:
C₉H₁₁BrN₂O Molecular Weight: 243.1004 g/mol |
MDL:
MFCD02681930 CAS-Nummer: 200064-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...