2-(2'-Chlorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-((2-Chlorobenzyl)oxy)phenyl)boronic acid

BB-8005-1G BB-8005-250M
COBBBB-8005-1GEA 204 EUR
COBBBB-8005-1G COBBBB-8005-250M
2-(2'-Chlorbenzyloxy)phenylboronsäure
2-(2'-Chlorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BClO₃
Molecular Weight: 262,496 g/mol
MDL: MFCD06200724
CAS-Nummer: 870777-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR