2-(3'-Chlorbenzyloxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-((3-Chlorobenzyl)oxy)phenyl)boronic acid
BB-8028-1G
BB-8028-250M
COBBBB-8028-1GEA
353
EUR
COBBBB-8028-1G
COBBBB-8028-250M
2-(3'-Chlorbenzyloxy)phenylboronsäure
2-(3'-Chlorbenzyloxy)phenylboronsäure
Formel:
C₁₃H₁₂BClO₃ Molecular Weight: 262,5000 g/mol |
MDL:
MFCD06411351 CAS-Nummer: 849062-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...