3-(2-Chlorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((2-Chlorobenzyl)oxy)phenyl)boronic acid

BB-8025-1G BB-8025-250M
COBBBB-8025-1GEA 79.5 EUR
COBBBB-8025-1G COBBBB-8025-250M
3-(2-Chlorbenzyloxy)phenylboronsäure
3-(2-Chlorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BClO₃
Molecular Weight: 262,5000 g/mol
MDL: MFCD05865198
CAS-Nummer: 845551-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR