3-(3-Chlorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((3-Chlorobenzyl)oxy)phenyl)boronic acid

BB-8088-1G BB-8088-250M
COBBBB-8088-1GEA 205 EUR
COBBBB-8088-1G COBBBB-8088-250M
3-(3-Chlorbenzyloxy)phenylboronsäure
3-(3-Chlorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BClO₃
Molecular Weight: 262,5 g/mol
MDL: MFCD06411352
CAS-Nummer: 849062-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR