3-(4-Chlorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8062-1G BB-8062-250M
COBBBB-8062-1GEA 204 EUR
COBBBB-8062-1G COBBBB-8062-250M
3-(4-Chlorbenzyloxy)phenylboronsäure
3-(4-Chlorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BClO₃
MDL: MFCD07369737
CAS-Nummer: 870778-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR