4-(4-Chlorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-((4-Chlorobenzyl)oxy)phenyl)boronic acid

BB-8038-1G BB-8038-250M
COBBBB-8038-1GEA 254 EUR
COBBBB-8038-1G COBBBB-8038-250M
4-(4-Chlorbenzyloxy)phenylboronsäure
4-(4-Chlorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BClO₃
Molecular Weight: 262,5000 g/mol
MDL: MFCD07784349
CAS-Nummer: 870778-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR