4-[(2,6-Difluorbenzyl)oxy]benzaldehyd

Lieferant: CHEMPUR

10-0013.1 10-0013.5
CHMP10-0013.1EA 0 EUR
CHMP10-0013.1 CHMP10-0013.5
4-[(2,6-Difluorbenzyl)oxy]benzaldehyd
4-[(2,6-Difluorbenzyl)oxy]benzaldehyd
Formel: C₁₄H₁₀F₂O₂
MDL: MFCD04114367
CAS-Nummer: 885269-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR