4-[(3,5-Difluorbenzyl)oxy]benzaldehyd

Lieferant: Thermo Scientific

SEW04748.250 SEW04748.1
MAYBSEW04748.250EA 0 EUR
MAYBSEW04748.250 MAYBSEW04748.1
4-[(3,5-Difluorbenzyl)oxy]benzaldehyd
4-[(3,5-Difluorbenzyl)oxy]benzaldehyd
Formel: C₁₄H₁₀F₂O₂
MDL: MFCD00278356
CAS-Nummer: 259132-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR