2,8-Dimethylchinolin-4(1H)-on

Lieferant: COMBI-BLOCKS

Synonyme: 2,8-Dimethylquinolin-4-ol, 2,8-Dimethyl-4-hydroxyquinoline

PY-1346-5G PY-1346-1G
COBBPY-1346-5GEA 126 EUR
COBBPY-1346-5G COBBPY-1346-1G
2,8-Dimethylchinolin-4(1H)-on
2,8-Dimethylchinolin-4(1H)-on
Formel: C₁₁H₁₁NO
Molecular Weight: 173.2111 g/mol
MDL: MFCD00100264
CAS-Nummer: 15644-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR