4,8-Dimethyl-2-chinolinol

Lieferant: BIONET RESEARCH

3N-501S.500 3N-501S.1 3N-501S.5 3N-501S.10
BIOT3N-501S.500EA 0 EUR
BIOT3N-501S.500 BIOT3N-501S.1 BIOT3N-501S.5 BIOT3N-501S.10
4,8-Dimethyl-2-chinolinol
4,8-Dimethyl-2-chinolinol
Formel: C₁₁H₁₁NO
MDL: MFCD00085676
CAS-Nummer: 5349-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR