Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate

HC-2500-5G HC-2500-1G HC-2500-250M
COBBHC-2500-5GEA 1170 EUR
COBBHC-2500-5G COBBHC-2500-1G COBBHC-2500-250M
Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255,0681 g/mol
MDL: MFCD07021501
CAS-Nummer: 886361-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR