3,8-Dibrom-6-methylimidazo[1,2-a]pyridin
Lieferant: COMBI-BLOCKS
HC-2126-5G
HC-2126-1G
COBBHC-2126-5GEA
303
EUR
COBBHC-2126-5G
COBBHC-2126-1G
3,8-Dibrom-6-methylimidazo[1,2-a]pyridin
3,8-Dibrom-6-methylimidazo[1,2-a]pyridin
Formel:
C₈H₆Br₂N₂ |
MDL:
MFCD11504900 CAS-Nummer: 1072944-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...