3,6-Dibrom-7-methylimidazo[1,2-a]pyridin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2158-1G HC-2158-5G
COBBHC-2158-1GEA 0 EUR
COBBHC-2158-1G COBBHC-2158-5G
3,6-Dibrom-7-methylimidazo[1,2-a]pyridin Hydrochlorid
3,6-Dibrom-7-methylimidazo[1,2-a]pyridin Hydrochlorid
Formel: C₈H₇Br₂ClN₂
MDL: MFCD11855860
CAS-Nummer: 1072944-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR