8-Brom-5-methylimidazo[1,2-a]pyridin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-6166-5G HC-6166-1G
COBBHC-6166-5GEA 1120 EUR
COBBHC-6166-5G COBBHC-6166-1G
8-Brom-5-methylimidazo[1,2-a]pyridin Hydrochlorid
8-Brom-5-methylimidazo[1,2-a]pyridin Hydrochlorid
Formel: C₈H₈BrClN₂
Molecular Weight: 247.5200 g/mol
MDL: MFCD09800975
CAS-Nummer: 957120-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR