5-Methylimidazo[1,2-a]pyridin-3-carbaldehyd
Lieferant: COMBI-BLOCKS
HC-6738-1G
HC-6738-5G
COBBHC-6738-1GEA
385
EUR
COBBHC-6738-1G
COBBHC-6738-5G
5-Methylimidazo[1,2-a]pyridin-3-carbaldehyd
5-Methylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel:
C₉H₈N₂O |
MDL:
MFCD11053823 CAS-Nummer: 178488-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...