6-Methylimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: COMBI-BLOCKS

HC-6663-5G HC-6663-1G
COBBHC-6663-5GEA 735 EUR
COBBHC-6663-5G COBBHC-6663-1G
6-Methylimidazo[1,2-a]pyridin-3-carbaldehyd
6-Methylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₉H₈N₂O
Molecular Weight: 160,1726 g/mol
MDL: MFCD09994344
CAS-Nummer: 933752-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR