6-Methyl-3-nitroimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-2005-5G HC-2005-1G
COBBHC-2005-5GEA 0 EUR
COBBHC-2005-5G COBBHC-2005-1G
6-Methyl-3-nitroimidazo[1,2-a]pyridin
6-Methyl-3-nitroimidazo[1,2-a]pyridin
Formel: C₈H₇N₃O₂
MDL: MFCD06496196
CAS-Nummer: 67625-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR