1H-Benzimidazol-5-carbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1H-Benzo[d]imidazole-5-carboxylic acid, Benzimidazole-5-carboxylic acid, 5-Benzimidazolecarboxylic acid

A15980.09 A15980.14 A15980.18 A15980.30
A15980.09EA 57 EUR
A15980.09 A15980.14 A15980.18 A15980.30
1H-Benzimidazol-5-carbonsäure ≥98%
1H-Benzimidazol-5-carbonsäure

Purity: 98% CAS No.: 15788-16-6 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 300° Beilstein Reference: 5686

Formel: C₈H₆N₂O₂
Molecular Weight: 162.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011555
CAS-Nummer: 15788-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR