4-(Isopropylsulfonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-(Isopropylsulphonyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2666-5G PN-2666-1G
COBBPN-2666-5GEA 1180 EUR
COBBPN-2666-5G COBBPN-2666-1G
4-(Isopropylsulfonyl)phenylboronsäurepinakolester
4-(Isopropylsulfonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₃BO₄S
Molecular Weight: 310.2200 g/mol
MDL: MFCD16618963
CAS-Nummer: 1256359-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR