2-(Methylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Methylsulphonyl)benzeneboronic acid, (2-(Methylsulphonyl)phenyl)boronic acid

BB-2643-1G BB-2643-25G BB-2643-5G
COBBBB-2643-1GEA 191 EUR
COBBBB-2643-1G COBBBB-2643-25G COBBBB-2643-5G
2-(Methylsulfonyl)phenylboronsäure
2-(Methylsulfonyl)phenylboronsäure
Formel: C₇H₉BO₄S
Molecular Weight: 200,02 g/mol
MDL: MFCD02179472
CAS-Nummer: 330804-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR