4-(Isopropylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Isopropylsulphonyl)benzeneboronic acid

BB-2666-5G BB-2666-1G
COBBBB-2666-5GEA 262 EUR
COBBBB-2666-5G COBBBB-2666-1G
4-(Isopropylsulfonyl)phenylboronsäure
4-(Isopropylsulfonyl)phenylboronsäure
Formel: C₉H₁₃BO₄S
Molecular Weight: 228,0700 g/mol
MDL: MFCD06659866
CAS-Nummer: 850567-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR