4-(Ethylsulfonyl)phenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(4-(Ethylsulphonyl)phenyl)boronic acid, 4-(Ethylsulphonyl)benzeneboronic acid
L17814.03
L17814.MD
L17814.03EA
116
EUR
L17814.03
L17814.MD
4-(Ethylsulfonyl)phenylboronsäure ≥98%
4-(Ethylsulfonyl)phenylboronsäure
Purity: 98+%
CAS No.: 352530-24-6
Molecular Formula: C8H11BO4S
Formula Weight: 214.05
Melting Point: 135-139°
MDL No.: MFCD02093076
Formel:
C₈H₁₁BO₄S Schmelzpunkt: 135…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093076 CAS-Nummer: 352530-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...