4-(Ethylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Ethylsulphonyl)phenyl)boronic acid, 4-(Ethylsulphonyl)benzeneboronic acid

BB-2664-5G BB-2664-1G
COBBBB-2664-5GEA 612 EUR
COBBBB-2664-5G COBBBB-2664-1G
4-(Ethylsulfonyl)phenylboronsäure
4-(Ethylsulfonyl)phenylboronsäure
Formel: C₈H₁₁BO₄S
Molecular Weight: 214,05 g/mol
MDL: MFCD02093076
CAS-Nummer: 352530-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR