1,1'-Bis(di-tert-butylphosphino)ferrocen ≥98%

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: 1,1'-Bis(di-t-butylphosphino)ferrocene, 1,1'-Bis[bis(1,1-dimethylethyl)phosphino]ferrocene, DTBPF

26-0150-500MG S26-0150.0002
STRE26-0150-500MGEA 149 EUR
STRE26-0150-500MG ABCRS26-0150.0002
1,1'-Bis(di-tert-butylphosphino)ferrocen ≥98%
1,1'-Bis(di-tert-butylphosphino)ferrocen

Phosphine

Formel: C₂₆H₄₄FeP₂
Molecular Weight: 474,42 g/mol
MDL: MFCD01630818
CAS-Nummer: 84680-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR