8-Brom-6-chlorimidazo[1,2-a]pyridin-3-carbaldehyd
Lieferant: COMBI-BLOCKS
HC-6733-1G
HC-6733-250M
COBBHC-6733-1GEA
0
EUR
COBBHC-6733-1G
COBBHC-6733-250M
8-Brom-6-chlorimidazo[1,2-a]pyridin-3-carbaldehyd
8-Brom-6-chlorimidazo[1,2-a]pyridin-3-carbaldehyd
Formel:
C₈H₄BrClN₂O |
MDL:
MFCD11504922 CAS-Nummer: 1093092-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...