Ethyl-8-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Lieferant: COMBI-BLOCKS
PY-7142-1G
PY-7142-5G
COBBPY-7142-1GEA
0
EUR
COBBPY-7142-1G
COBBPY-7142-5G
Ethyl-8-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-8-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Formel:
C₁₀H₈BrClN₂O₂ |
MDL:
MFCD09607953 CAS-Nummer: 951884-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...