8-Brom-6-chlorimidazo[1,2-a]pyridin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-6164-1G
HC-6164-5G
COBBHC-6164-1GEA
0
EUR
COBBHC-6164-1G
COBBHC-6164-5G
8-Brom-6-chlorimidazo[1,2-a]pyridin Hydrochlorid
8-Brom-6-chlorimidazo[1,2-a]pyridin Hydrochlorid
Formel:
C₇H₅BrCl₂N₂ |
MDL:
MFCD09800974 CAS-Nummer: 957120-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...