8-Brom-6-chlor-3-nitroimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-2124-5G HC-2124-1G
COBBHC-2124-5GEA 227 EUR
COBBHC-2124-5G COBBHC-2124-1G
8-Brom-6-chlor-3-nitroimidazo[1,2-a]pyridin
8-Brom-6-chlor-3-nitroimidazo[1,2-a]pyridin
Formel: C₇H₃BrClN₃O₂
MDL: MFCD11504898
CAS-Nummer: 1072944-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR