3-Brom-7-methoxyimidazo[1,2-a]pyridin-8-carbonitril
Lieferant: COMBI-BLOCKS
HC-2086-5G
HC-2086-1G
COBBHC-2086-5GEA
0
EUR
COBBHC-2086-5G
COBBHC-2086-1G
3-Brom-7-methoxyimidazo[1,2-a]pyridin-8-carbonitril
3-Brom-7-methoxyimidazo[1,2-a]pyridin-8-carbonitril
Formel:
C₉H₆BrN₃O |
MDL:
MFCD11504889 CAS-Nummer: 1072944-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...