8-Hydroxyimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: COMBI-BLOCKS

HC-6735-5G HC-6735-1G
COBBHC-6735-5GEA 884 EUR
COBBHC-6735-5G COBBHC-6735-1G
8-Hydroxyimidazo[1,2-a]pyridin-3-carbaldehyd
8-Hydroxyimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₈H₆N₂O₂
MDL: MFCD10699667
CAS-Nummer: 1033202-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR