7-Methoxyimidazo[1,2-a]pyridin-8-carbonitril

Lieferant: COMBI-BLOCKS

HC-6315-5G HC-6315-1G
COBBHC-6315-5GEA 375 EUR
COBBHC-6315-5G COBBHC-6315-1G
7-Methoxyimidazo[1,2-a]pyridin-8-carbonitril
7-Methoxyimidazo[1,2-a]pyridin-8-carbonitril
Formel: C₉H₇N₃O
MDL: MFCD09801017
CAS-Nummer: 834869-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR