Methylimidazo[1,2-a]pyridin-5-carboxylat
Lieferant: COMBI-BLOCKS
HC-2502-1G
HC-2502-250M
COBBHC-2502-1GEA
246
EUR
COBBHC-2502-1G
COBBHC-2502-250M
Methylimidazo[1,2-a]pyridin-5-carboxylat
Methylimidazo[1,2-a]pyridin-5-carboxylat
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,1700 g/mol |
MDL:
MFCD12026020 CAS-Nummer: 88047-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...