Methylimidazo[1,2-a]pyridin-6-carboxylat

Lieferant: BIONET RESEARCH

7X-0857.5 7X-0857.500 7X-0857.1 7X-0857.10
BIOT7X-0857.5EA 0 EUR
BIOT7X-0857.5 BIOT7X-0857.500 BIOT7X-0857.1 BIOT7X-0857.10
Methylimidazo[1,2-a]pyridin-6-carboxylat
Methylimidazo[1,2-a]pyridin-6-carboxylat
Formel: C₉H₈N₂O₂
MDL: MFCD06200984
CAS-Nummer: 136117-69-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR