Ethylimidazo[1,2-a]pyridin-7-carboxylat

Lieferant: COMBI-BLOCKS

HI-1237-5G HI-1237-1G
COBBHI-1237-5GEA 451 EUR
COBBHI-1237-5G COBBHI-1237-1G
Ethylimidazo[1,2-a]pyridin-7-carboxylat
Ethylimidazo[1,2-a]pyridin-7-carboxylat
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,1986 g/mol
MDL: MFCD08436046
CAS-Nummer: 372147-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR