Methylimidazo[1,2-a]pyridin-6-carboxylat Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-6274-5G
HC-6274-1G
COBBHC-6274-5GEA
0
EUR
COBBHC-6274-5G
COBBHC-6274-1G
Methylimidazo[1,2-a]pyridin-6-carboxylat Hydrochlorid
Methylimidazo[1,2-a]pyridin-6-carboxylat Hydrochlorid
Formel:
C₉H₉ClN₂O₂ |
MDL:
MFCD09801003 CAS-Nummer: 957120-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...