Ethylimidazo[1,2-a]pyridin-7-carboxylat Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-6194-1G HC-6194-5G
COBBHC-6194-1GEA 0 EUR
COBBHC-6194-1G COBBHC-6194-5G
Ethylimidazo[1,2-a]pyridin-7-carboxylat Hydrochlorid
Ethylimidazo[1,2-a]pyridin-7-carboxylat Hydrochlorid
Formel: C₁₀H₁₁ClN₂O₂
MDL: MFCD09800986
CAS-Nummer: 957120-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR