Imidazo[1,2-a]pyridin-6-carbonsäure

Lieferant: COMBI-BLOCKS

HI-1240-5G HI-1240-1G
COBBHI-1240-5GEA 0 EUR
COBBHI-1240-5G COBBHI-1240-1G
Imidazo[1,2-a]pyridin-6-carbonsäure
Imidazo[1,2-a]pyridin-6-carbonsäure
Formel: C₈H₆N₂O₂
MDL: MFCD07021498
CAS-Nummer: 139022-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR