Imidazo[1,2-a]pyridin-7-carbonsäure
Lieferant: COMBI-BLOCKS
HI-1363-5G
HI-1363-1G
COBBHI-1363-5GEA
0
EUR
COBBHI-1363-5G
COBBHI-1363-1G
Imidazo[1,2-a]pyridin-7-carbonsäure
Imidazo[1,2-a]pyridin-7-carbonsäure
Formel:
C₈H₆N₂O₂ |
MDL:
MFCD09834781 CAS-Nummer: 648423-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...