Imidazo[1,2-a]pyridin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-6990-5G HC-6990-1G
COBBHC-6990-5GEA 124 EUR
COBBHC-6990-5G COBBHC-6990-1G
Imidazo[1,2-a]pyridin Hydrochlorid
Imidazo[1,2-a]pyridin Hydrochlorid
Formel: C₇H₇ClN₂
MDL: MFCD09954953
CAS-Nummer: 34167-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR