Imidazo[1,2-a]pyridin-8-ol

Lieferant: BIONET RESEARCH

Synonyme: 8-Hydroxyimidazo[1,2-a]pyridine

8X-0841.1 8X-0841.500
BIOT8X-0841.1EA 0 EUR
BIOT8X-0841.1 BIOT8X-0841.500
Imidazo[1,2-a]pyridin-8-ol
Imidazo[1,2-a]pyridin-8-ol
Formel: C₇H₆N₂O
Molecular Weight: 134.1400 g/mol
MDL: MFCD06496222
CAS-Nummer: 69214-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR