Imidazo[1,2-a]pyridin-8-ol Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 8-Hydroxyimidazo[1,2-a]pyridine hydrochloride

HC-6116-1G HC-6116-5G
COBBHC-6116-1GEA 175 EUR
COBBHC-6116-1G COBBHC-6116-5G
Imidazo[1,2-a]pyridin-8-ol Hydrochlorid
Imidazo[1,2-a]pyridin-8-ol Hydrochlorid
Formel: C₇H₇ClN₂O
MDL: MFCD09800942
CAS-Nummer: 100592-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR