Ethyl-5-methylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: COMBI-BLOCKS

PY-7134-25G PY-7134-1G PY-7134-5G
COBBPY-7134-25GEA 358 EUR
COBBPY-7134-25G COBBPY-7134-1G COBBPY-7134-5G
Ethyl-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,230 g/mol
MDL: MFCD02186029
CAS-Nummer: 67625-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR