Ethyl-6-methylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: COMBI-BLOCKS

PY-7135-1G PY-7135-5G
COBBPY-7135-1GEA 0 EUR
COBBPY-7135-1G COBBPY-7135-5G
Ethyl-6-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-6-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₂N₂O₂
MDL: MFCD02325396
CAS-Nummer: 70705-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR