Ethyl-7-methylimidazo[1,2-a]pyridin-2-carboxylat
Lieferant: COMBI-BLOCKS
PY-7136-1G
PY-7136-5G
COBBPY-7136-1GEA
103
EUR
COBBPY-7136-1G
COBBPY-7136-5G
Ethyl-7-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-7-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel:
C₁₁H₁₂N₂O₂ |
MDL:
MFCD02187526 CAS-Nummer: 70705-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...